Skip to Content
Merck
All Photos(2)

Documents

GBL247882

Grace Bio-Labs ProPlate® microarray system tray set

size 2 wells, with Delrin® snap clips

Synonym(s):

incubation chambers, microarray hybridization chambers, slide chambers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.15

material

black silicone (Acrylic tray)
rectangular wells

sterility

non-sterile

feature

wells 2

packaging

pack of 1 ea

manufacturer/tradename

Grace Bio-Labs 247882

size

2 wells

well volume

350 μL

color

none

Looking for similar products? Visit Product Comparison Guide

General description

ProPlate re-usable, multi-well chambers form removable, leak-proof wells on virtually any slide surface without the use of adhesive.
Designs feature ANSI-SLAS compliant well spacing to facilitate multichannel pipetting.
Multiple slides insert easily into frames having a standard microtiter plate footprint for automated processing.

Legal Information

Delrin is a registered trademark of E. I. du Pont de Nemours and Company
ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service