Skip to Content
Merck
All Photos(1)

Key Documents

GA4

85062502, Application: Fusion partner

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Sheep Heterohybridoma

ImmunoDon

Sheep lymph node

Myeloma

NS1

Reactivity

Non Ig secreting

technique(s)

cell culture | mammalian: suitable

isotype

None

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NS1 x Sheep lymph node

Cell Line Description

This HAT sensitive line can be used as a fusion partner in the production of heterohybridomas.

Immunogen

Bovine red blood cells

Application

Fusion partner

Culture Medium

RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 10% Horse Serum (HS)

Subculture Routine

Maintain cultures between 2-6x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service