Skip to Content
Merck
All Photos(1)

Documents

AC-0352A

Sigma-Aldrich

Claudin-4 (EP417) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

shipped in

wet ice

storage temp.

2-8°C

visualization

(membrane)

General description

Claudin-4 is one of twenty-seven integral membrane tight junction proteins and specializes in paracellular sodium transport. Although claudin proteins are primarily known for their cell adhesion function, Claudin-4 binds to zonula occludens proteins and interacts with numerous signaling pathways. Claudin-4 is detectable in low amounts in the normal tissues of the lung, kidney, breast, prostate, bladder and gastrointestinal tract. It is a highly specific and sensitive membranous marker for most epithelial neoplasms. Absence of expression in normal mesothelium and mesothelioma provides utility in the differential diagnosis between carcinoma from mesothelioma with 99% specificity. In breast tumors, claudin-4 levels are significantly higher in ER-negative and high grade tumors when compared to ER-positive and low grade tumors. High levels of Claudin-4 expression are associated with later stages of progression and reduced patient survival for most types of epithelial neoplasms. Conversely, its expression is downregulated in gastric and high-grade bladder cancers, and is significantly lower in lymph node metastasis and esophageal tumors with distant metastasis. Low Claudin-4 expression is associated with a poor prognosis in esophageal squamous cell carcinoma.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1%BSA and &< 0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact IVDorder@ABCAM.com

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service