Skip to Content
Merck
All Photos(1)

Key Documents

320099

Sigma-Aldrich

Acetic acid

ACS reagent, ≥99.7%

Synonym(s):

Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

ACS reagent

vapor density

2.07 (vs air)

vapor pressure

11.4 mmHg ( 20 °C)

Assay

≥99.7%

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

impurities

Cr2O72- reducers, passes test
MnO4- reducers, passes test
≤0.0004 meq/g Titr. base
≤0.01% acetic anhydride

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.371 (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 ppm
sulfate (SO42-): ≤1 ppm

cation traces

Fe: ≤0.2 ppm
heavy metals (as Pb): ≤0.5 ppm

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sukant Khurana et al.
Chemical senses, 38(4), 315-323 (2013-02-01)
We studied complete dose-response curves for 53 odorants in the third instar larvae of Drosophila melanogaster. All odorants, except one, elicited an attraction response. Some odorants also elicited a decrease from their peak response at higher concentrations. This concentration-dependent decrease
John E P Syka et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(26), 9528-9533 (2004-06-24)
Peptide sequence analysis using a combination of gas-phase ion/ion chemistry and tandem mass spectrometry (MS/MS) is demonstrated. Singly charged anthracene anions transfer an electron to multiply protonated peptides in a radio frequency quadrupole linear ion trap (QLT) and induce fragmentation
Eagleson M.
Concise Encyclopedia Chemistry, 5-5 (1994)
Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue
Katharine Trenholme et al.
Antimicrobial agents and chemotherapy, 58(7), 3666-3678 (2014-04-16)
Therapies to prevent transmission of malaria parasites to the mosquito vector are a vital part of the global malaria elimination agenda. Primaquine is currently the only drug with such activity; however, its use is limited by side effects. The development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service