Skip to Content
Merck
All Photos(1)

Documents

NIST1128

Titanium-base alloy

NIST® SRM® 1128, 15V - 3Al - 3Cr - 3Sn

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

General description

The Standard Reference Material, titanium-base alloy, is in the form of a disk with dimensions of 35 mm (1 3/8 in) in diameter and 19 mm (3/4 in) in thickness. For more information, please refer to the SDS and COA.
SRM 1128_cert
SRM 1128_SDS

Application

SRM1128 is intended for use in optical emission and x-ray fluorescence spectrometric methods of analysis.

Other Notes

Certified for the analytes listed below.
Vanadium (V), Aluminium (Al), Tin (Sn), Chromium (Cr), Iron (Fe), Carbon (C)

Reference/Informational value is provided for the following analytes.
Boron (B), Copper (Cu), Magnesium (Mg), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Niobium (Nb), Nitrogen (N)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service