G0400100
Glyceryl trinitrate solution
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Glycerol trinitrate solution, 1,2,3-Propanetriol 1,2,3-trinitrate solution
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
glyceryl trinitrate
form
solution
contains
~99% Ethanol
manufacturer/tradename
EDQM
concentration
≤1%
application(s)
pharmaceutical (small molecule)
format
neat
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
53.6 °F - closed cup
Flash Point(C)
12 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service