Skip to Content
Merck
All Photos(1)

Key Documents

95615

Supelco

Xylenol Orange

metal indicator (for complexometry)

Synonym(s):

o-Cresolsulfonphthalein-3′-3″-bis(methyliminodiacetic acid sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H32N2O13S
CAS Number:
Molecular Weight:
672.66
EC Number:
MDL number:
UNSPSC Code:
12161500

grade

metal indicator (for complexometry)

SMILES string

[Na+].Cc1cc(cc(CN(CC(O)=O)CC(O)=O)c1O)\C(=C2\C=C(C)C(=O)C(CN(CC(O)=O)CC(O)=O)=C2)c3ccccc3S([O-])(=O)=O

Other Notes

Metallochromic ligand employed in chelation ion chromatography of trace metal ions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier Díaz-Castro et al.
Pediatrics, 134(2), 257-264 (2014-07-16)
Clamping and cutting of the umbilical cord is the most prevalent of all operations, but the optimal timing of cord clamping is controversial, with different timings offering advantages and disadvantages. This study, for the first time, compares the influence of
Pui Kit Suen et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 32(8), 997-1005 (2014-05-02)
Sclerostin is a negative regulator of bone formation. Sclerostin monoclonal antibody (Scl-Ab) treatment promoted bone healing in various animal models. To further evaluate the healing efficiency of Scl-Ab in osteotomy healing, we investigated the time course effects of systemic administration
B. Paull et al.
Trends in Analytical Chemistry, 18, 107-107 (1999)
Tracy A Brennan et al.
Disease models & mechanisms, 7(5), 583-592 (2014-03-15)
A major medical challenge in the elderly is osteoporosis and the high risk of fracture. Telomere dysfunction is a cause of cellular senescence and telomere shortening, which occurs with age in cells from most human tissues, including bone. Telomere defects
Rhandy M Eman et al.
Stem cells and development, 23(24), 2950-2958 (2014-07-19)
Bone defect healing is highly dependent on the simultaneous stimulation of osteogenesis and vascularization. In bone regenerative strategies, combined seeding of multipotent stromal cells (MSCs) and endothelial progenitor cells (EPCs) proves their mutual stimulatory effects. Here, we investigated whether stromal-cell-derived

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service