Skip to Content
Merck
All Photos(1)

Key Documents

66042

Supelco

Withanoside V

analytical standard

Synonym(s):

(1α,3β,22R)-3-[(6-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]-1,22-dihydroxy-ergosta-5,24-dien-26-oic acid δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H62O14
CAS Number:
Molecular Weight:
766.91
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C[C@H]([C@H]1CC(C)=C(C)C(=O)O1)[C@H]2CC[C@H]3[C@@H]4CC=C5C[C@H](C[C@H](O)[C@]5(C)[C@H]4CC[C@]23C)O[C@@H]6O[C@H](CO[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)[C@@H](O)[C@H](O)[C@H]6O

InChI

1S/C40H62O14/c1-17-12-26(52-36(49)18(17)2)19(3)23-8-9-24-22-7-6-20-13-21(14-29(42)40(20,5)25(22)10-11-39(23,24)4)51-38-35(48)33(46)31(44)28(54-38)16-50-37-34(47)32(45)30(43)27(15-41)53-37/h6,19,21-35,37-38,41-48H,7-16H2,1-5H3/t19-,21+,22-,23+,24-,25-,26+,27+,28+,29-,30+,31+,32-,33-,34+,35+,37+,38+,39+,40-/m0/s1

InChI key

ZBLWKSUMHLVXAM-KFJRISAASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shreya Dubey et al.
PloS one, 13(11), e0200053-e0200053 (2018-11-08)
Withania somnifera (Ashwagandha) is an efficient medicinal plant known in Ayurveda and Chinese medicine since ancient times, whose extracts are consumed orally as food supplement or as a health tonic owing to its several restorative properties for various CNS disorders

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service