Skip to Content
Merck
All Photos(1)

Key Documents

33852

Supelco

Decanal 2,4-dinitrophenylhydrazone

analytical standard

Synonym(s):

Caprinaldehyde-2,4-DNPH, Decyl aldehyde-2,4-DNPH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
336.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC/CHN)

shelf life

limited shelf life, expiry date on the label

feature

standard type calibration

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

100-104 °C

application(s)

environmental

format

neat

SMILES string

CCCCCCCCC\C=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C16H24N4O4/c1-2-3-4-5-6-7-8-9-12-17-18-15-11-10-14(19(21)22)13-16(15)20(23)24/h10-13,18H,2-9H2,1H3/b17-12+

InChI key

WVTWCMFTBSTXFK-SFQUDFHCSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service