Skip to Content
Merck
All Photos(1)

Documents

33698

Supelco

Profoxydim lithium salt

PESTANAL®, analytical standard

Synonym(s):

Clefoxydim lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31ClLiNO4S
CAS Number:
Molecular Weight:
471.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[Li+].CCC\C(=N/OCC(C)Oc1ccc(Cl)cc1)C2=C([O-])CC(CC2=O)C3CCCSC3

InChI

1S/C24H32ClNO4S.Li/c1-3-5-21(26-29-14-16(2)30-20-9-7-19(25)8-10-20)24-22(27)12-18(13-23(24)28)17-6-4-11-31-15-17;/h7-10,16-18,27H,3-6,11-15H2,1-2H3;/q;+1/p-1/b26-21+;

InChI key

LRGTYGSKTGIXOL-HCPJHKKFSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Profoxydim is a pesticide used for the control of crabgrass, yellow bristle grass and barnyard grass in seeded and transplanted rice.

Application

Profoxydim lithium salt may be used as an analytical reference standard for the determination of profoxydim in rice grain samples and rice paddy systems using matrix solid-phase dispersion (MSPD) extraction followed by high-performance liquid chromatography-diode array detection (HPLC-DAD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Herbicide Classes in Development, 397(6), 2181-2190 (2012)
A multi-residue method for pesticide residue analysis in rice grains using matrix solid-phase dispersion extraction and high-performance liquid chromatography-diode array detection.
Tsochatzis DE, et al.
Analytical and Bioanalytical Chemistry, 397(6), 2181-2190 (2010)
E D Tsochatzis et al.
Chemosphere, 91(7), 1049-1057 (2013-03-20)
Rice cultivation relies on pesticide applications to ensure high yields. However, the regular use of pesticides seriously affects the quality of neighboring surface water systems. Thus complete knowledge of the environmental fate and dissipation of pesticides in the paddy rice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service