Skip to Content
Merck
All Photos(1)

Documents

03818

Sigma-Aldrich

Ethylene glycol monodecyl ether

≥97.0% (GC)

Synonym(s):

2-(Decyloxy)ethanol, C10E1, Decylglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9OCH2CH2OH
CAS Number:
Molecular Weight:
202.33
Beilstein:
1849710
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

Assay

≥97.0% (GC)

refractive index

n20/D 1.441

density

0.875 g/mL at 20 °C (lit.)

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCOCCO

Other Notes

Non-ionic surfactant which effectively releases histamine from rat peritoneal mast cells

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of surfactants upon rat peritoneal mast cells in vitro.
C Prottey et al.
Food and cosmetics toxicology, 14(5), 425-430 (1976-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service