Skip to Content
Merck
All Photos(1)

Documents

28-4120

Sigma-Aldrich

Sodium silicate solution

CP, liquid

Synonym(s):

Sodium trisilicate solution, Water glass

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2O(SiO2)x · xH2O
UNSPSC Code:
12352302

grade

CP

form

liquid

availability

available only in Japan

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service