Skip to Content
Merck
All Photos(1)

Key Documents

15-1470

Sigma-Aldrich

Iron(III) sulfate hydrate

JIS special grade, 60.0-80.0% anhydrous basis

Synonym(s):

Ferric sulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
399.88 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
JIS special grade
form:
solid

grade

JIS special grade

Assay

60.0-80.0% anhydrous basis

form

solid

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

storage temp.

15-25°C

SMILES string

[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Fe.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6

InChI key

RUTXIHLAWFEWGM-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gal Frenkel et al.
The Journal of clinical pediatric dentistry, 37(2), 137-141 (2012-01-01)
To compare the long-term clinical and radiographic outcomes of pulpotomies in primary molars performed with white or gray Mineral Trioxide Aggregate (MTA) in combination with ferric sulfate (FS), when one package of MTA is used for multiple treatments. Sixty eight
Y Yilmaz et al.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry, 12(2), 117-122 (2011-06-15)
The aim of this study was to compare the local anaesthetic efficacy of articaine HCl and prilocaine HCl during an operative procedure after their administration by either mandibular nerve block or maxillary infiltration. The study was a double-blind clinical study
Oksana Velgosová et al.
Waste management (New York, N.Y.), 33(2), 456-461 (2012-11-08)
The paper is concerned with biohydrometallurgical methods of cadmium recovery from spent Ni-Cd batteries. Cd leaching efficiency from electrode material in different media (H(2)SO(4) and Fe(2)(SO(4))(3) solutions), at different Fe(III) concentrations and using the bacteria Acidithiobacillus ferrooxidans were investigated. The
Sudip Bhattacharyya et al.
Free radical research, 46(10), 1296-1307 (2012-07-24)
In the present study we investigated the beneficial role of glycine in iron (FeSO₄) induced oxidative damage in murine hepatocytes. Exposure of hepatocytes to 20 μM FeSO₄ for 3 hours enhanced reactive oxygen species (ROS) generation and induced alteration in
J Uusi-Kämppä et al.
Journal of environmental quality, 41(2), 420-426 (2012-03-01)
Permanent grass vegetation on sloping soils is an option to protect fields from erosion, but decaying grass may liberate considerable amounts of dissolved reactive P (DRP) in springtime runoff. We studied the effects of freezing and thawing of grassed soil

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service