RIE156322380
Rieke Metals, Inc.
2-(3-Iodobenzoyl)oxazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(3-iodophenyl)(1,3-oxazol-2-yl)methanone
Inchi Code
1S/C10H6INO2/c11-8-3-1-2-7(6-8)9(13)10-12-4-5-14-10/h1-6H
InChI key
DWDQFOOKAGAKFR-UHFFFAOYSA-N
Purity
97%
Country of Origin
US
Physical Form
Light yellow solid
Product Link
https://riekemetals.com/products-services/catnum/5572-21
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.