Skip to Content
Merck
All Photos(1)

Documents

ST1681

Sigma-Aldrich

Anti-SIGIRR Mouse mAb (3H8-2G3)

liquid, clone 3H8-2G3, Calbiochem®

Synonym(s):

Anti-Single Immunoglobulin and Toll-Interleukin 1 Receptor Domain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3H8-2G3, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgM

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIGIRR(59307)

General description

Mouse monoclonal antibody supplied as ascites. Recognizes the ~32 kDa SIGIRR protein.
Recognizes the ~32 kDa SIGIRR protein in HepG2 cells.
This Anti-SIGIRR Mouse mAb (3H8-2G3) is validated for use in Immunoblotting for the detection of SIGIRR.

Immunogen

Full-length, recombinant, human SIGIRR (aa 1-411), expressed as a GST fusion protein

Application

Immunoblotting (1-5 µg/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

Undiluted ascites

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
HepG2 cells

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service