Skip to Content
Merck
All Photos(1)

Documents

MABF2248

Sigma-Aldrich

Anti-EBV LMP1 Antibody, clone S12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

clone

S12
monoclonal

purified by

using protein G

species reactivity

mouse, virus

concentration

(Please refer to lot specific datasheet.)

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

General description

A fusion protein (pKH548) consisting of 189 amino acids from the C-terminus region of LMP1 coupled to beta-galactosidase.

Specificity

Clone S12 is a mouse monoclonal antibody that specifically detects Latent membrane protein 1 (LMP1) in Epstein-Barr virus transformed cells.

Application

Immunocytochemistry Analysis: A representative lot detected EBV LMP1 in Immunocytochemistry applications (Xu, J., et. al. (2002). J Virol. 76(8):4080-6).

Flow Cytometry Analysis: A representative lot detected EBV LMP1 in Flow Cytometry applications (Mann, K.P., et. al. (1985). J Virol. 55(3):710-20).

Western Blotting Analysis: A representative lot detected EBV LMP1 in Western Blotting applications (Choi, I.K., et. al. (2018). Proc Natl Acad Sci USA. 115(4):E686-E695; Young, L.S., et. al. (1988). J Gen Virol. 69 ( Pt 5):1051-65; Mann, K.P., et. al. (1985). J Virol. 55(3):710-20).

Immunofluorescence Analysis: A representative lot detected EBV LMP1 in Immunofluorescence applications (Mann, K.P., et. al. (1985). J Virol. 55(3):710-20).

Immunohistochemistry Analysis: A representative lot detected EBV LMP1 in Immunohistochemistry applications (Brink, A.A., et. al. (1997). J Clin Pathol. 50(11):911-8; Khabir, A., et. al. (2005). Virol J. 2:39).

Immunoprecipitation Analysis: A representative lot immunoprecipitated EBV LMP1 in Immunoprecipitation applications (Young, L.S., et. al. (1988). J Gen Virol. 69 ( Pt 5):1051-65).

Quality

Evaluated by Western Blotting in Raji cell lysates.

Western Blotting Analysis: 1 µg/mL of this antibody detected LMP1 in lysates from Raji cells.

Physical properties

~59 kDa observed; 41.98 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Virus transformed human cells.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service