MAB2626
Anti-Endosialin Antibody, clone B1/35
clone B1/35, from mouse
Synonym(s):
CD248 molecule, endosialin, CD164 sialomucin-like 1, endosialin, CD248 antigen, endosialin, Tumor endothelial marker 1, CD248 antigen
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
B1/35, monoclonal
species reactivity
human
technique(s)
flow cytometry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CD248(57124)
Related Categories
General description
Immunogen
Application
Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in IP on AG1523 human fibroblasts (MacFadyen, J. et al. (2005). FEBS. 579(12):2569-2575).
Immunofluorescent Analysis: A previous lot was used by an independent laboratory in IF to detect Endosialin in human rheumatoid synovium, tonsil, salivary gland and normal breast tissues (MacFadyen, J. et al. (2005). FEBS. 579(12):2569-2575).
Quality
Western Blot Analysis: A 1:2,500 dilution of this antibody detected Endosialin in 10 µg of AG1523 cell lysate.
Target description
Physical form
Analysis Note
AG1523 cell lysate
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service