Skip to Content
Merck
All Photos(1)

Key Documents

FF-1L-BK

Millipore

Anti-Fyb (clone SpA264LBg1), FFMU

Anti-Fyb from the clone SpA264LBg1, Human IgM Antibody, Intermediate for further manufacturing use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

biological source

human

Quality Level

antibody form

unpurified

clone

SpA264LBg1, monoclonal

form

liquid

packaging

pkg of 1 L

concentration

2-10 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

6.7-7.7

isotype

IgM

shipped in

ambient

storage temp.

2-8°C

General description

Standardized, concentrated cell culture supernatant which has been diafiltered with phosphate buffered saline.; For the formulation of Anti-s blood typing reagents or devices.; Contains 0.1% (w/v) Sodium Azide; Shelf Life: currently being established.

Application

Antibody intermediate for further manufacturing use
Production of an in vitro blood typing product or device

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service