Skip to Content
Merck

Skip To

DDMISC0010

Direct Detect® Spotting Tray

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41116129
NACRES:
NB.22
eCl@ss:
32160606


manufacturer/tradename

Direct Detect®

technique(s)

protein quantification: suitable

shipped in

ambient

General description

The Direct DetectTM Spotting Tray is designed to hold 5 Assay-free Cards for rapid spotting.

Legal Information

DIRECT DETECT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DDAC00010Z743362Z743360
description

-

description

50 Membrane cards used for protein quantitation in the Direct Detect® Infrared Spectrometer.

description

blue, L × W 2.0 in. × 0.75 in., pkg of 500 roll of

description

blue, L × W 1.50 in. × 0.50 in., pkg of 750 roll of

shipped in

ambient

shipped in

ambient

shipped in

-

shipped in

-

manufacturer/tradename

Direct Detect®

manufacturer/tradename

Direct Detect®

manufacturer/tradename

Diversified Biotech DTCR-7000-B

manufacturer/tradename

Diversified Biotech DTCR-2000-B

technique(s)

protein quantification: suitable

technique(s)

protein quantification: suitable

technique(s)

cryopreservation: suitable

technique(s)

cryopreservation: suitable




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters