Skip to Content
Merck
All Photos(2)

Key Documents

ABE2984

Sigma-Aldrich

Anti-Phospho-RXRα/RXRA (Ser56)

Synonym(s):

Nuclear receptor subfamily 2 group B member 1, Retinoic acid receptor RXR-alpha, Retinoid X receptor alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

mol wt

calculated mol wt 50.81 kDa
observed mol wt ~75-80 kDa

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine, rat, mouse

packaging

antibody small pack of 100

technique(s)

dot blot: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... RXRA(6256)

Specificity

This rabbit polyclonal antibody detects RXR phosphorylated on serine 56.

Immunogen

KLH-conjugated linear peptide corresponding to 12 amino acids from the surrounding phosphoserine 56 from the N-terminal region of human Retinoic acid receptor (RXR ).

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HeLa cells in mitotic phase.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RXR phosphorylated on serine 56 in lysate from HeLa cells in mitotic phase.

Tested Applications

Immunofluorescence Analysis: A representative lot detected RXR in Immunofluorescence application (Xie, G., et al. (2021). STAR Protocols. 2, :1-15).

Western Blotting Analysis: A representative lot detected RXR in Western Blotting application (Xie, G., et al. (2020). Development Cell. 55, : 707-722).

Immunoprecipitation Analysis: A representative lot immunoprecipitated RXR in Immunoprecipitation application (Xie, G., et al. (2020). Development Cell. 55, : 707-722).

Immunocytochemistry Analysis: A representative lot detected RXR in Immunocytochemistry application (Xie, G., et al. (2020). Development Cell. 55, : 707-722).

Flow Cytometry Analysis: A representative lot detected RXR in Flow Cytometry application (Xie, G., et al. (2020). Development Cell. 55, : 707-722).

Dot Blot: A representative lot detected RXR in Dot Blot application (Xie, G., et al. (2020). Development Cell. 55, : 707-722).

Flow Cytometry Analysis: 1.0 µg of this antibody detected RXR in HeLa cells overnight serum-starved and treated with Nocodazole 100 ng/ml for 8 hours .

Immunohistochemistry Applications: A representative lot detected RXR in Immunohistochemistry application (Xie, G., et al. (2020). Development Cell. 55, : 707-722).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Retinoic acid receptor RXR-alpha (UniProt: P19793; also known as Nuclear receptor subfamily 2 group B member 1, Retinoid X receptor alpha, RXR ) is encoded by the RXRA (also known as NR2B1) gene (Gene ID: 6256) in human. Retinoid X receptor alpha (RXR ), a nuclear receptor of transcription factor, controls various physiological and pathological pathways including cellular growth, proliferation, differentiation, and apoptosis. RXR-alpha is composed of three domains: a modulating N-terminal domain (AF1 domain), a DNA-binding domain, and a C-terminal ligand-binding domain (AF2 domain). RXR-alpha protein is highly expressed in liver and its expression has also been reported in lung fibroblasts, monocytes, kidney, and brain. RXR is phosphorylated on serine and threonine residues mainly at its N-terminal A/B domain by cyclin-dependent kinase 1 (Cdk1) at the onset of mitosis, triggering its translocation to the centrosome, where phosphorylated-RXR (p-RXR ) interacts with polo-like kinase 1 (PLK1) through its N-terminal A/B domain by a unique mechanism. The interaction promotes PLK1 activation, centrosome maturation, and mitotic progression. Studies show that Cdk1 phosphorylation of Ser56 and Ser70 mediates the interaction between p-RXR and PLK1 during mitosis. Importantly, Ser56 and Ser70 and their surrounding amino acids are highly conserved across different species, suggesting their evolutionarily conserved role in regulating RXR activity. An RXR ligand XS060, which specifically inhibits p-RXR /PLK1 interaction but not RXR heterodimerization, promotes mitotic arrest and catastrophe in a tumor-specific manner. Levels of p-RXR are abnormally elevated in cancer cell lines, during carcinogenesis in animals, and in clinical tumor tissues. (Ref.: Xie, G., et al. (2020). Development Cell. 55(6); 707-722; Xie, G., et al. (2021). STAR Protocols. 2(3); 1-15; Zeng, Z., et al. (2015). Cancer Res. 75(10); 2049-2060.)

Physical form

Purified rabbit polyclonal antibody in PBS without azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service