Skip to Content
Merck
All Photos(1)

Documents

8.14087

Sigma-Aldrich

Hexadecyltrimethylammonium chloride

(50% solution in 2-propanol/water 3:2) for synthesis

Synonym(s):

Cetyltrimethylammonium chloride solution, Trimethylhexadecylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107
NACRES:
NA.22

Quality Level

form

liquid

potency

500-2000 mg/kg LD50, oral (Rat)

pH

5-7 (20 °C, 10 g/L in H2O)

transition temp

flash point 15 °C

density

0.89 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (argentometric): 49.0 - 52.0 %
Density (d 20 °C/ 4 °C): 0.887 - 0.892

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1C - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F

Flash Point(C)

15 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service