Skip to Content
Merck
All Photos(1)

Documents

1.11157

Supelco

Chlorine and pH Test Kit Refill pack, colorimetric

for use with MQUANT®

Synonym(s):

Chlorine and pH Visual Test Kit Refill for Chlorine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Chlorine and pH Test Refill pack Reagents for chlorine (Cl₂-1, Cl₂-2, Cl₂-3), for use with MQUANT®

feature

refill

Quality Level

specific analyte(s)

chlorine

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

When the reagents of a MQuant® test (formally MColortest®) are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of test kits, making it a particularly economical and ecological system.
This refill pack contains all the reagents for the MQuant® test kits 1.11174 for the colometric evaluation of chlorine.
for 111174 MQuant®

Application

  • Green synthesis and characterization of binary, ternary, and quaternary Ti/MMO anodes for chlorine and oxygen evolution reactions.: This study presents the development of titanium mixed metal oxide (Ti/MMO) anodes for chlorine and oxygen evolution, crucial for electrolysis processes in water treatment (Abdel-Aziz et al., 2024).
  • Experimental study on mine water purification mechanism for broken coal and rock masses in the underground reservoir of ecologically vulnerable mining area.: Focuses on the purification processes of mine water in fragile ecological zones, using methods including chlorination, to manage pollution and improve water quality (Wang et al., 2024).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for free Chlorine according to DIN EN ISO 7393

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service