Skip to Content
Merck
All Photos(1)

Documents

1.07185

Sigma-Aldrich

Pepsin

(from porcine gastric mucosa) 0.7 FIP-U/mg for biochemistry EC 3.4.23.1

Synonym(s):

Pepsin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
35000
MDL number:
UNSPSC Code:
12352204
EC Index Number:
232-629-3
NACRES:
NA.21

pH

4.0-5.0 (20 °C, 10 g/L in H2O)

solubility

10 g/L

bulk density

380‑420 kg/m3

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

Application

Pepsin has been used in the preparation of simulated gastric fluid to perform in vitro gastric digestion.

Biochem/physiol Actions

Pepsin is a component of gastric juice and is associated with protein digestion. It is an endopeptidase that catalyzes the degradation of proteins into peptides and amino acids for small intestinal absorption. Pepsin is released as a zymogen (pepsinogen) by the chief cells of the gastric lining. It gets activated by the acidic environment created by the hydrochloric acid (HCl) of gastric juice. Pepsin is active at pH 1.5 to 2, in the stomach. It is involved in the conversion of food bolus into chyme. It is associated with laryngopharyngeal reflux (LPR) indicated by chronic cough, mild dysphagia, hoarseness, and non-productive throat clearing.

Analysis Note

Appearance: fine, slightly yellow powder
Activity/FIP (Hemoglobin; pH 1.6; 25 °C): ≥ 0.70 FIP-U/mg
pH-value (1 %; water): 4.0 - 5.0
Water (according to Karl Fischer): ≤ 6.0 %

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service