ENAH961CE66E
Enamine
2-hydroxy-3-methyl-1,4-dihydronaphthalene-1,4-dione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
168-173
Iupac Name
2-hydroxy-3-methylnaphthoquinone
Inchi Code
1S/C11H8O3/c1-6-9(12)7-4-2-3-5-8(7)11(14)10(6)13/h2-6H,1H3
InChI key
XGQIWAFRNOGWHB-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-1273208
Melting Point
168-173
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-1273208
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.