Skip to Content
Merck
All Photos(1)

Key Documents

ENAH961CE55B

Enamine

3-amino-4-phenylbutanamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
214.69
MDL number:

mp

214-216

Iupac Name

3-amino-4-phenylbutanamide hydrochloride

Inchi Code

1S/C10H14N2O.ClH/c11-9(7-10(12)13)6-8-4-2-1-3-5-8;/h1-5,9H,6-7,11H2,(H2,12,13);1H

InChI key

LIIYHDNLCSGEMR-UHFFFAOYSA-N

Salt Data

Cl

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-8785647

Melting Point

214-216

MSDS

https://www.enaminestore.com/catalog/EN300-8785647


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library