Skip to Content
Merck

ENA387772288

Enamine

5,6-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
$67.20

$67.20


Estimated to ship onMay 23, 2025


To order products, please contact your local dealer.

Select a Size

Change View
1 AMP
$67.20

About This Item

CAS Number:
Molecular Weight:
229.71
MDL number:

$67.20


Estimated to ship onMay 23, 2025


To order products, please contact your local dealer.

Iupac Name

5,6-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

Inchi Code

1S/C11H15NO2.ClH/c1-13-10-4-3-8-7-12-6-5-9(8)11(10)14-2;/h3-4,12H,5-7H2,1-2H3;1H

InChI key

QPBAENLXKAWWOO-UHFFFAOYSA-N

storage temp.

4

Salt Data

Cl

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-119688

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-119688


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library