Skip to Content
Merck
All Photos(1)

Documents

791661P

Avanti

CER10(d9)

N-palmitoyl(d9) dihydrosphingosine, powder

Synonym(s):

N-palmitoyl(d9) sphinganine; 16:0-d9 DH Ceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H60D9NO3
CAS Number:
Molecular Weight:
548.97
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (791661P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791661P

shipped in

dry ice

storage temp.

−20°C

General description

N-palmitoyl(d9) dihydrosphingosine is a deuterated form of N-palmitoyl dihydrosphingosine (N16DS18). N16DS18 is a N-acyl dihydrosphingosine (NDS) present in human hair. NDS acyl chains comprise 2-24 carbons.

Application

N-palmitoyl(d9) dihydrosphingosine CER10(d9) has been used as a deuterium-labeled ceramide internal standard in lipid extraction from human and mouse stratum corneum (SC) samples for liquid chromatography tandem-mass spectrometry (LC–MS/MS) analysis.

Biochem/physiol Actions

Dihydrosphingosine provides protection to the skin by acting as a lipid barrier.

Packaging

5 mL Amber Glass Screw Cap Vial (791661P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshinori Masukawa et al.
Journal of chromatography. A, 1127(1-2), 52-59 (2006-06-20)
An analytical method for highly sensitive determination of four N-acyl dihydrosphingosines (NDSs) of all ceramides (CERs) in human hair, such as N-palmitoyl dihydrosphingosine (N16DS18), N-stearoyl dihydrosphingosine (N18DS18), N-lignocerol dihydrosphingosine (N24DS18) and N-nervonoyl dihydrosphingosine (N24:1DS18), has been developed using electrospray ionization
Barbora Školová et al.
Biochimica et biophysica acta. Biomembranes, 1859(5), 824-834 (2017-01-23)
Ceramides based on phytosphingosine, sphingosine and dihydrosphingosine are essential constituents of the skin lipid barrier that protects the body from excessive water loss. The roles of the individual ceramide subclasses in regulating skin permeability and the reasons for C4-hydroxylation of
Momoko Kawana et al.
Journal of lipid research, 61(6), 884-895 (2020-04-09)
Ceramides are the predominant lipids in the stratum corneum (SC) and are crucial components for normal skin barrier function. Although the composition of various ceramide classes in the human SC has been reported, that in mice is still unknown, despite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service