ATEH9614FF3E
AstaTech, Inc.
TERT-BUTYL ((2-CHLOROPYRIMIDIN-5-YL)METHYL)CARBAMATE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
tert-butyl ((2-chloropyrimidin-5-yl)methyl)carbamate
Inchi Code
1S/C10H14ClN3O2/c1-10(2,3)16-9(15)14-6-7-4-12-8(11)13-5-7/h4-5H,6H2,1-3H3,(H,14,15)
InChI key
VUFBZTGHKQVWPB-UHFFFAOYSA-N
Purity
95%
Country of Origin
CN
Physical Form
White to Yellow Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=165441
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=W19322
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.