ATE959439771
AstaTech, Inc.
(R)-3-METHYL-2-(1-OXOISOINDOLIN-2-YL)BUTANOIC ACID
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(2R)-3-methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
Inchi Code
1S/C13H15NO3/c1-8(2)11(13(16)17)14-7-9-5-3-4-6-10(9)12(14)15/h3-6,8,11H,7H2,1-2H3,(H,16,17)/t11-/m1/s1
InChI key
JULAETYRRYJIAR-LLVKDONJSA-N
Purity
95%
Physical Form
Pale-yellow to Yellow-brown Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=110430
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=56630
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.