Skip to Content
Merck
All Photos(1)

Key Documents

ATE458969610

AstaTech, Inc.

5-BROMO-PYRROLO[2,1-F][1,2,4]TRIAZINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
198.02
MDL number:

Iupac Name

5-bromopyrrolo[2,1-f][1,2,4]triazine

Inchi Code

1S/C6H4BrN3/c7-5-1-2-10-6(5)3-8-4-9-10/h1-4H

InChI key

UHGYITTVLZNRNT-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=69251

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=P13049


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library