Skip to Content
Merck
All Photos(1)

Key Documents

ATE410297401

AstaTech, Inc.

9-(TERT-BUTYLDIMETHYLSILYL)OXYMETHYL-2-AMINOFLUORENE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
325.53
MDL number:

Iupac Name

9-(((tert-butyldimethylsilyl)oxy)methyl)-9H-fluoren-2-amine

Inchi Code

1S/C20H27NOSi/c1-20(2,3)23(4,5)22-13-19-16-9-7-6-8-15(16)17-11-10-14(21)12-18(17)19/h6-12,19H,13,21H2,1-5H3

InChI key

NUKGMRZJRUHHMM-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

98%

Shipping Temperature

Normal

Physical Form

Yellow to Brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=22951

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=SC5065


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library