Skip to Content
Merck
All Photos(1)

Key Documents

APO455829748

Apollo Scientific Ltd

2-[4-(4,5-Dihydro-1,3-thiazol-2-yl)phenoxy]acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
218.28
MDL number:

bp

83-85

Iupac Name

[4-(4,5-dihydro-1,3-thiazol-2-yl)phenoxy]acetonitrile

Inchi Code

1S/C11H10N2OS/c12-5-7-14-10-3-1-9(2-4-10)11-13-6-8-15-11/h1-4H,6-8H2

InChI key

GQEVWXYAFGXFEJ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/2-4-45-dihydro-13-thiazol-2-ylphenoxyacetonitrile

Boiling point

83-85

Pictograms

Skull and crossbones

Signal Word

Danger

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-4-45-dihydro-13-thiazol-2-ylphenoxyacetonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library