Skip to Content
Merck
All Photos(1)

Key Documents

W315400

Sigma-Aldrich

2-Ethyl-5(6)-methylpyrazine

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
FEMA Number:
3154
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
N/A

biological source

synthetic

grade

Kosher

Assay

≥98%

form

liquid

refractive index

n20/D 1.502-1.505 (lit.)

bp

57 °C/10 mmHg (lit.)

density

0.964 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

potato

SMILES string

CCc1cnc(C)cn1.CCc2cncc(C)n2

InChI

1S/2C7H10N2/c1-3-7-5-8-6(2)4-9-7;1-3-7-5-8-4-6(2)9-7/h2*4-5H,3H2,1-2H3

InChI key

MHVIQJDXHDKUAM-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.0 °F - closed cup

Flash Point(C)

58.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service