Documents
THS03134
ThalesNano CatCart® catalyst cartridge system, 70 mm L
nickel sponge, 1% Mo
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
UNSPSC Code:
41103817
NACRES:
NB.51
packaging
pack of 1 ea
manufacturer/tradename
ThalesNano THS 03134
diam.
4 mm
Related Categories
General description
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Legal Information
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.