Key Documents
TEM-3752
Sigma-Aldrich
HPM100 Leica adaptor for High Pressure Freezing with CryoCapsule
About This Item
UNSPSC Code:
41111761
General description
Adaptor that loads CryoCapsule, and allow High Pressure Freezing Using HPM100 machine for correlative light and electron microscopy (CLEM). For step-by-step usage please see: https://www.sciencedirect.com/science/article/pii/B9780128010754000124
Choose from one of the most recent versions:
Certificates of Analysis (COA)