Skip to Content
Merck
All Photos(1)

Documents

DWKFDF124A

Azlon Clear PMP Volumetric Flask with PPE Stopper

class A, tolerance, capacity 100 mL

Synonym(s):

FDF124A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

material

round clear PMP flask

feature

class A tolerance

packaging

pkg of 1 ea

manufacturer/tradename

Azlon FDF124A

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

Application

For the accurate preparation of solutions where glass alternatives would not be suitable

Features and Benefits

Excellent clarity and calibrated to ISO 1042

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service