Skip to Content
Merck
All Photos(1)

Documents

CF0005

Sigma-Aldrich

Alcohol Togni-(4-Br-PhOCF2F2)-reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H14BrF4IO2
CAS Number:
MDL number:
UNSPSC Code:
12352200

form

solid

reaction suitability

reaction type: C-C Bond Formation

storage temp.

−20°C

Application

Alcohol Togni-(4-Br-PhOCF2CF2)-reagent is an electrophilic hypervalent iodine-fluoroalkyl reagent introducing the 4-bromophenoxytetrafluoroethyl fragment. The aromatic bromide can be further subjected to Pd-catalyzed cross-coupling reactions to build molecular complexity.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service