Skip to Content
Merck
All Photos(1)

Documents

BR781840

BRAND® microplate BRANDplates®, lipoGrade

size 96 wells, size 330 μL, round bottom, transparent

Synonym(s):

BRANDplates®, Immunology, Microplates, lipoGrade

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

colorless wells
polystyrene
round bottom

sterility

non-sterile

feature

lid: no
skirt (U-bottom)

packaging

pack of 100 ea

manufacturer/tradename

BRAND 781840

size

330 μL
96 wells

working volume

330 μL

color

transparent

Looking for similar products? Visit Product Comparison Guide

General description

For the immobilization of hydrophobic moleculesStrongly hydrophobic (lipophilic), for immobilization of biomolecules with predominantly hydrophobic regions.An alternative to the immunoGrade surface for the immobilization of molecules, such as lipoproteins or peptides.Specially suited for liquid phase assays when the reaction component should stay in solution. The majority of hydrophilic biomolecules are not immobilized on this surface.PS, non-sterile.BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens

Features and Benefits

• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service