BR114316
BRAND® test tube, graduated, glass
tube capacity 20 mL, without stopper, with spout
Synonym(s):
Test tubes, Tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass tube
colorless tube
round bottom
sterility
non-sterile
feature
graduations
without stopper, with spout
packaging
pack of 10 ea
manufacturer/tradename
BRAND 114316
tube capacity
20 mL
tube diam. × L
17 mm × 180 mm
Looking for similar products? Visit Product Comparison Guide
General description
Graduation of approx. volume and inscriptions in high contrast white enamel. Wall thickness approx. 1.2 mm.
Features and Benefits
- BRAND® graduated test tubes are versatile to fit most laboratory applications.
- Manufactured from clear borosilicate glass for easy viewing of contents.
- Includes a polypropylene cap for optimal sample recovery.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service