PEG Mn 4000-6000 g/mol (by NMR) PEG average Mn 5000 PLGA Mn 13500-16500 g/mol (by NMR) PLGA average Mn 15000
color
white
shipped in
dry ice
storage temp.
−20°C
Application
Amine poly(ethylene glycol)-block-poly(lactide-co-glycolide) is an amine-functionalized, biocompatible, amphiphilic block copolymer. Due to its self-assembling properties, this material can be used in the formation of nanoparticles for drug delivery. Additionally, due to the amine-functionalization of the PEG block, this material can be used to rapidly covalently conjugate biomolecules, API, or targeting ligands. Potential uses include the targeted and/or controlled release of cancer drugs, anti-inflammatory drugs, antibiotics, or anaesthetic agents.
Journal of medicinal chemistry, 60(10), 4510-4516 (2017-05-05)
Polymeric nanoparticles (PNPs) may efficiently deliver in vivo therapeutics to tumors when conjugated to specific targeting agents. Gint4.T aptamer specifically recognizes platelet-derived growth factor receptor β and can cross the blood-brain barrier (BBB). We synthesized Gint4.T-conjugated PNPs able of high
Journal of biomedical materials research. Part B, Applied biomaterials, 105(6), 1692-1716 (2016-04-22)
Poly (lactic-co-glycolic acid) (PLGA) copolymers have been broadly used in controlled drug release applications. Because these polymers are biodegradable, they provide an attractive option for drug delivery vehicles. There are a variety of material, processing, and physiological factors that impact
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.