Skip to Content
Merck
All Photos(1)

Key Documents

774774

Sigma-Aldrich

1-Bromo-2,4,6-tricyclopentylbenzene

95%

Synonym(s):

2,4,6-Tricyclopentylbromobenzene, 2,4,6-Tricyclopentylphenyl bromide, 2-Bromo-1,3,5-tricyclopentylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H29Br
CAS Number:
Molecular Weight:
361.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

95%

density

1.183 g/mL at 25 °C

storage temp.

−70°C

SMILES string

Brc1c(cc(cc1C2CCCC2)C3CCCC3)C4CCCC4

InChI

1S/C21H29Br/c22-21-19(16-9-3-4-10-16)13-18(15-7-1-2-8-15)14-20(21)17-11-5-6-12-17/h13-17H,1-12H2

InChI key

QRLTUCJNCQBOBR-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F

Flash Point(C)

47 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service