Skip to Content
Merck
All Photos(1)

Key Documents

765775

Sigma-Aldrich

1,1,1-Trifluoro-3-methyl-2-butanol

95%

Synonym(s):

3-Methyl-1,1,1-trifluoro-2-butanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9F3O
CAS Number:
Molecular Weight:
142.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.354

bp

98-104 °C

density

1.149 g/mL at 25 °C

functional group

fluoro
hydroxyl

storage temp.

2-8°C

SMILES string

CC(C)C(O)C(F)(F)F

InChI

1S/C5H9F3O/c1-3(2)4(9)5(6,7)8/h3-4,9H,1-2H3

InChI key

FPIWJHHQCNHQIC-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service