Skip to Content
Merck
All Photos(1)

Key Documents

750212

Sigma-Aldrich

(3-Ammoniopropyl)diisopropylphosphonium bis(tetrafluoroborate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H24B2F8NP
Molecular Weight:
350.88
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

300-310 °C

functional group

phosphine

SMILES string

F[B-](F)(F)F.F[B-](F)(F)F.CC(C)[PH+](CCC[NH3+])C(C)C

InChI

1S/C9H22NP.2BF4/c1-8(2)11(9(3)4)7-5-6-10;2*2-1(3,4)5/h8-9H,5-7,10H2,1-4H3;;/q;2*-1/p+2

InChI key

XOFTUYXVVIJLRD-UHFFFAOYSA-P

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service