Skip to Content
Merck
All Photos(1)

Key Documents

744735

Sigma-Aldrich

1,3-Dihydro-1-hydroxy-3,3-dimethyl-1,2-benziodoxole

98.0-102.0% (AT)

Synonym(s):

1-Hydroxy-1,3-dihydro-3,3-dimethyl-1,2-benziodoxole, NSC 312854

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$495.00

$495.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$495.00

About This Item

Empirical Formula (Hill Notation):
C9H11IO2
CAS Number:
Molecular Weight:
278.09
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$495.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98.0-102.0% (AT)

form

powder

functional group

iodo

storage temp.

2-8°C

SMILES string

OI1OC(C)(C)C2=CC=CC=C21

InChI

1S/C9H11IO2/c1-9(2)7-5-3-4-6-8(7)10(11)12-9/h3-6,11H,1-2H3

InChI key

LYUULLIYJCLGFU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Organoiodinane reagents for phosphate cleavage: experimental and computational studies.
Moss, R.A., et al.
Journal of the American Chemical Society, 111, 250-258 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service