Skip to Content
Merck
All Photos(1)

Key Documents

730858

Sigma-Aldrich

Diallyl-d10-amine

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD2=CDCD2)2NH
Molecular Weight:
107.22
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

liquid

bp

111-112 °C (lit.)

mp

-88 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

mass shift

M+10

SMILES string

[2H]\C([2H])=C(/[2H])C([2H])([2H])NC([2H])([2H])\C([2H])=C(\[2H])[2H]

InChI

1S/C6H11N/c1-3-5-7-6-4-2/h3-4,7H,1-2,5-6H2/i1D2,2D2,3D,4D,5D2,6D2

InChI key

DYUWTXWIYMHBQS-URTNXKOFSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service