Skip to Content
Merck
All Photos(1)

Key Documents

680729

Sigma-Aldrich

4-Hydroxy-2-cyclopentenone

96%

Synonym(s):

4-Hydroxy-2-cyclopenten-1-one, 4-Oxo-2-cyclopenten-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10
MDL number:
UNSPSC Code:
12352100

Assay

96%

contains

0.5% BHT as stabilizer

refractive index

n20/D 1.5078

density

1.1623 g/mL at 25 °C

SMILES string

OC1CC(=O)C=C1

Looking for similar products? Visit Product Comparison Guide

Application

A building block for drug synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Asymmetry, 15, 2807-2809 (2004)
Tetrahedron, 53 (6), 1983-2004 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service