Skip to Content
Merck
All Photos(1)

Key Documents

521647

Sigma-Aldrich

Ethyl 6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate

98%

Synonym(s):

3-Ethoxycarbonyl-6,7,8-trifluoro-1,4-hydro-4-oxoquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8F3NO3
CAS Number:
Molecular Weight:
271.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

275 °C (dec.) (lit.)

SMILES string

CCOC(=O)C1=CNc2c(F)c(F)c(F)cc2C1=O

InChI

1S/C12H8F3NO3/c1-2-19-12(18)6-4-16-10-5(11(6)17)3-7(13)8(14)9(10)15/h3-4H,2H2,1H3,(H,16,17)

InChI key

ONQDAESGZUODFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate is also referred to as 3-ethoxycarbonyl-6,7,8-trifluoro-1,4-hydro-4-oxoquinoline.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Anaya-Gonzalez et al.
Free radical biology & medicine, 141, 150-158 (2019-06-14)
Phototoxic effects of 6,8 dihalogenated quinolones confers to this type of molecules a potential property as photochemotherapeutic agents. Two photodehalogenation processes seem to be involved in the remarkable photoinduced cellular damage. In this context, a new 6,8 dihalogenated quinolone 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service