512567
Bis(ethylcyclopentadienyl)tungsten(IV) dihydride
95%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H20W
CAS Number:
Molecular Weight:
372.15
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
Assay
95%
reaction suitability
core: tungsten
reagent type: catalyst
mp
48-52 °C (lit.)
SMILES string
[H][W][H].CC[C]1[CH][CH][CH][CH]1.CC[C]2[CH][CH][CH][CH]2
InChI
1S/2C7H9.W.2H/c2*1-2-7-5-3-4-6-7;;;/h2*3-6H,2H2,1H3;;;
InChI key
XHAKKDCRMPIPFF-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service