Skip to Content
Merck
All Photos(1)

Key Documents

406163

Sigma-Aldrich

1-Benzyl-2,3-isopropylidene-rac-glycerol

99%

Synonym(s):

(±)-4-Benzyloxymethyl-2,2-dimethyl-1,3-dioxolane, DL-α,β-Isopropylideneglycerol-γ-benzyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O3
CAS Number:
Molecular Weight:
222.28
Beilstein:
82961
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.494 (lit.)

bp

89-91 °C/0.1 mmHg (lit.)

density

1.051 g/mL at 25 °C (lit.)

SMILES string

CC1(C)OCC(COCc2ccccc2)O1

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service