Skip to Content
Merck
All Photos(1)

Key Documents

400998

Sigma-Aldrich

Cerium(III) bromide

99%

Synonym(s):

Cerium tribromide, Cerous bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeBr3
CAS Number:
Molecular Weight:
379.83
EC Number:
MDL number:
UNSPSC Code:
12352302

Assay

99%

mp

730 °C (lit.)
730-732 °C (lit.)

SMILES string

Br[Ce](Br)Br

InChI

1S/3BrH.Ce/h3*1H;/q;;;+3/p-3

InChI key

MOOUSOJAOQPDEH-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service